Product Details

SNP ID
rs149161953
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:15493946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGAGCCCACTGCTCAAAGATGTC[A/G]TCCAGGGTCTCAACGTACCAGGAGC
Phenotype
MIM: 602234 MIM: 609444
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP9 PubMed Links

Gene Details

Gene
CASP9
Gene Name
caspase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001229.4 899 Silent Mutation GAC,GAT D368D NP_001220.2
NM_001278054.1 899 Silent Mutation GAC,GAT D218D NP_001264983.1
NM_032996.3 899 Silent Mutation GAC,GAT D285D NP_127463.2
XM_005246014.2 899 Silent Mutation GAC,GAT D285D XP_005246071.1
XM_011542273.2 899 Silent Mutation GAC,GAT D368D XP_011540575.1
Gene
CELA2B
Gene Name
chymotrypsin like elastase family member 2B
There are no transcripts associated with this gene.

View Full Product Details