Product Details
- SNP ID
-
rs149191301
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:159826899 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTTGCCTGCCTCTCCAGAGAAAG[A/T]TGGTCATGAGGCCCCTGTGGAGTCT
- Phenotype
-
MIM: 606620
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf204
PubMed Links
Gene Details
- Gene
- C1orf204
- Gene Name
- chromosome 1 open reading frame 204
There are no transcripts associated with this gene.
- Gene
- SLAMF8
- Gene Name
- SLAM family member 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020125.2 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
NP_064510.1 |
XM_005245345.2 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
XP_005245402.1 |
XM_011509775.1 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
XP_011508077.1 |
XM_011509776.2 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
XP_011508078.1 |
XM_011509777.2 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
XP_011508079.1 |
XM_011509778.2 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
XP_011508080.1 |
XM_011509779.2 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
XP_011508081.1 |
XM_011509780.2 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
XP_011508082.2 |
XM_017001844.1 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
XP_016857333.1 |
XM_017001845.1 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
XP_016857334.1 |
XM_017001846.1 |
250 |
Missense Mutation |
ATG,TTG |
M1L |
XP_016857335.1 |
View Full Product Details