Product Details

SNP ID
rs149331570
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:161260463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCCCTTTTCACTGAGGTCTAGG[A/G]TTCATACTACTCCTCCCACCCCGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCP4L1 PubMed Links

Gene Details

Gene
PCP4L1
Gene Name
Purkinje cell protein 4 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102566.1 Intron NP_001096036.1
XM_017002154.1 Intron XP_016857643.1

View Full Product Details