Product Details

SNP ID
rs149447916
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:231347802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAATATTTTATGTTCCAGACCCT[C/G]CTGCATGAAATGATACATGCCTATT
Phenotype
MIM: 615283 MIM: 616086
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXOC8 PubMed Links

Gene Details

Gene
EXOC8
Gene Name
exocyst complex component 8
There are no transcripts associated with this gene.

Gene
SPRTN
Gene Name
SprT-like N-terminal domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010984.3 775 Silent Mutation CTC,CTG L109L NP_001010984.1
NM_001261462.2 775 Intron NP_001248391.1
NM_032018.6 775 Silent Mutation CTC,CTG L109L NP_114407.3
XM_006711818.2 775 Intron XP_006711881.1
XM_011544289.2 775 Intron XP_011542591.1

View Full Product Details