Product Details

SNP ID
rs149482751
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:200552547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAGCCCCCATTATGAAACACAG[A/G]TAACAGGGTAAACTACTGTGCCAGC
Phenotype
MIM: 611279
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF14 PubMed Links

Gene Details

Gene
KIF14
Gene Name
kinesin family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305792.1 5539 UTR 3 NP_001292721.1
NM_014875.2 5539 UTR 3 NP_055690.1
XM_011510231.2 5539 UTR 3 XP_011508533.1
XM_011510232.2 5539 UTR 3 XP_011508534.1
XM_011510233.2 5539 UTR 3 XP_011508535.1
XM_011510235.2 5539 UTR 3 XP_011508537.1
XM_011510236.1 5539 UTR 3 XP_011508538.1
XM_017003005.1 5539 UTR 3 XP_016858494.1
XM_017003006.1 5539 UTR 3 XP_016858495.1
XM_017003007.1 5539 UTR 3 XP_016858496.1

View Full Product Details