Product Details

SNP ID
rs149597334
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:93151693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGCAACAGGTGAATAAGTACATA[C/T]AGAAACAGAACTCCAATGCAAAGTA
Phenotype
MIM: 616876
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMED5 PubMed Links

Gene Details

Gene
TMED5
Gene Name
transmembrane p24 trafficking protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167830.1 4163 UTR 3 NP_001161302.1
NM_016040.4 4163 UTR 3 NP_057124.3

View Full Product Details