Product Details

SNP ID
rs149675198
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:151048380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACCCTGAGCCAGTGGTCCACAC[C/T]TGGGTCTACCCCGAGCCGGTGGCCG
Phenotype
MIM: 611275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BNIPL PubMed Links

Gene Details

Gene
BNIPL
Gene Name
BCL2 interacting protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159642.1 598 Intron NP_001153114.1
NM_138278.3 598 Intron NP_612122.2
XM_011509236.1 598 Intron XP_011507538.1
XM_017000420.1 598 Intron XP_016855909.1
Gene
C1orf56
Gene Name
chromosome 1 open reading frame 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017860.3 598 Missense Mutation CCT,CTT P178L NP_060330.2
Gene
CDC42SE1
Gene Name
CDC42 small effector 1
There are no transcripts associated with this gene.

View Full Product Details