Product Details

SNP ID
rs149696758
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:156311061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTATGGCCATTGTTCCACACC[A/C]GTCATGGCCTTGGATTTTTCTGTCA
Phenotype
MIM: 600114
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCT3 PubMed Links

Gene Details

Gene
CCT3
Gene Name
chaperonin containing TCP1 subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008800.2 1407 Silent Mutation ACG,ACT T392T NP_001008800.1
NM_005998.4 1407 Silent Mutation ACG,ACT T430T NP_005989.3

View Full Product Details