Product Details
- SNP ID
-
rs149911255
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
21
- Location
-
Chr.1:220058849 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCCAAATGAAACTTTCCTTTAAG[A/G]AACAAGTGCATTTTTAATAGATTCT
- Phenotype
-
MIM: 604053
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BPNT1
PubMed Links
Gene Details
- Gene
- BPNT1
- Gene Name
- 3'(2'), 5'-bisphosphate nucleotidase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286149.1 |
1098 |
Missense Mutation |
CCT,TCT |
P253S |
NP_001273078.1 |
NM_001286150.1 |
1098 |
Missense Mutation |
CCT,TCT |
P272S |
NP_001273079.1 |
NM_001286151.1 |
1098 |
Missense Mutation |
CCT,TCT |
P253S |
NP_001273080.1 |
NM_006085.5 |
1098 |
Missense Mutation |
CCT,TCT |
P308S |
NP_006076.4 |
XM_005272998.2 |
1098 |
Missense Mutation |
CCT,TCT |
P323S |
XP_005273055.1 |
XM_005272999.4 |
1098 |
Missense Mutation |
CCT,TCT |
P323S |
XP_005273056.1 |
XM_005273000.4 |
1098 |
Missense Mutation |
CCT,TCT |
P308S |
XP_005273057.1 |
XM_005273004.2 |
1098 |
Missense Mutation |
CCT,TCT |
P242S |
XP_005273061.1 |
XM_005273005.4 |
1098 |
Missense Mutation |
CCT,TCT |
P242S |
XP_005273062.1 |
XM_006711114.3 |
1098 |
Missense Mutation |
CCT,TCT |
P287S |
XP_006711177.1 |
XM_011509063.2 |
1098 |
Missense Mutation |
CCT,TCT |
P268S |
XP_011507365.1 |
XM_011509064.2 |
1098 |
Missense Mutation |
CCT,TCT |
P232S |
XP_011507366.1 |
XM_011509065.2 |
1098 |
Intron |
|
|
XP_011507367.1 |
XM_017000043.1 |
1098 |
Missense Mutation |
CCT,TCT |
P272S |
XP_016855532.1 |
XM_017000044.1 |
1098 |
Missense Mutation |
CCT,TCT |
P217S |
XP_016855533.1 |
XM_017000045.1 |
1098 |
Intron |
|
|
XP_016855534.1 |
XM_017000046.1 |
1098 |
Missense Mutation |
CCT,TCT |
P187S |
XP_016855535.1 |
View Full Product Details