Product Details

SNP ID
rs150172728
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:93562283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAGGAGGAGGTTCCAATTTACG[C/G]GAGAGGGCAGTTAAAATCTGGTTGA
Phenotype
MIM: 604704 MIM: 608848
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BCAR3 PubMed Links

Gene Details

Gene
BCAR3
Gene Name
breast cancer anti-estrogen resistance 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261408.1 2327 Silent Mutation TCC,TCG S812S NP_001248337.1
NM_001261409.1 2327 Silent Mutation TCC,TCG S812S NP_001248338.1
NM_001261410.1 2327 Silent Mutation TCC,TCG S721S NP_001248339.1
NM_001308251.1 2327 Silent Mutation TCC,TCG S488S NP_001295180.1
NM_003567.3 2327 Silent Mutation TCC,TCG S812S NP_003558.1
XM_011542251.2 2327 Silent Mutation TCC,TCG S592S XP_011540553.1
XM_017002480.1 2327 Silent Mutation TCC,TCG S812S XP_016857969.1
Gene
FNBP1L
Gene Name
formin binding protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024948.2 2327 Intron NP_001020119.1
NM_001164473.2 2327 Intron NP_001157945.1
NM_017737.4 2327 Intron NP_060207.2
XM_011541625.2 2327 Intron XP_011539927.1
XM_017001533.1 2327 Intron XP_016857022.1

View Full Product Details