Product Details

SNP ID
rs150254518
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:54757985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCCTCTGTGATGTGGTCGTACA[A/G]CTGCCCTATGAGCTCGGAGGCCGCC
Phenotype
MIM: 612036
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARS2 PubMed Links

Gene Details

Gene
PARS2
Gene Name
prolyl-tRNA synthetase 2, mitochondrial (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152268.3 1299 Silent Mutation CTG,TTG L393L NP_689481.2

View Full Product Details