Product Details

SNP ID
rs150405344
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:19683325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGCTGCCTGGCGAGGCTCCTC[A/G]GAGGGCAGGCAGCCTGGGGCCAGCA
Phenotype
MIM: 601109
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTR6 PubMed Links

Gene Details

Gene
HTR6
Gene Name
5-hydroxytryptamine receptor 6
There are no transcripts associated with this gene.

Gene
TMCO4
Gene Name
transmembrane and coiled-coil domains 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181719.4 2337 Silent Mutation NP_859070.3
XM_005245822.4 2337 Silent Mutation XP_005245879.1
XM_005245824.4 2337 Silent Mutation XP_005245881.1
XM_005245825.4 2337 Silent Mutation XP_005245882.1
XM_005245827.3 2337 Silent Mutation XP_005245884.1
XM_006710551.3 2337 Silent Mutation XP_006710614.1
XM_011541179.2 2337 Silent Mutation XP_011539481.1
XM_011541180.2 2337 Silent Mutation XP_011539482.1
XM_011541181.1 2337 Silent Mutation XP_011539483.1
XM_011541185.2 2337 Silent Mutation XP_011539487.1
XM_011541186.2 2337 Silent Mutation XP_011539488.1
XM_017000913.1 2337 Silent Mutation XP_016856402.1
XM_017000914.1 2337 Silent Mutation XP_016856403.1
XM_017000915.1 2337 Silent Mutation XP_016856404.1
XM_017000916.1 2337 Silent Mutation XP_016856405.1
XM_017000917.1 2337 Silent Mutation XP_016856406.1
XM_017000918.1 2337 Silent Mutation XP_016856407.1
XM_017000919.1 2337 Silent Mutation XP_016856408.1
XM_017000920.1 2337 Silent Mutation XP_016856409.1
XM_017000921.1 2337 Silent Mutation XP_016856410.1
XM_017000922.1 2337 Silent Mutation XP_016856411.1

View Full Product Details