Product Details
- SNP ID
-
rs150756783
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:91262571 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAATTTGATAAAGAAATATCCGA[C/T]ATCTCATGTGATGAAACAAAACTAA
- Phenotype
-
MIM: 615684
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HFM1
PubMed Links
Gene Details
- Gene
- HFM1
- Gene Name
- HFM1, ATP dependent DNA helicase homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001017975.4 |
3142 |
Missense Mutation |
ATA,ATG |
I1332M |
NP_001017975.4 |
XM_011540849.1 |
3142 |
Missense Mutation |
ATA,ATG |
I1332M |
XP_011539151.1 |
XM_011540850.2 |
3142 |
Missense Mutation |
ATA,ATG |
I1332M |
XP_011539152.1 |
XM_011540851.1 |
3142 |
Missense Mutation |
ATA,ATG |
I1332M |
XP_011539153.1 |
XM_011540852.2 |
3142 |
Missense Mutation |
ATA,ATG |
I1332M |
XP_011539154.1 |
XM_011540855.1 |
3142 |
Missense Mutation |
ATA,ATG |
I1290M |
XP_011539157.1 |
XM_011540857.1 |
3142 |
Missense Mutation |
ATA,ATG |
I1191M |
XP_011539159.1 |
XM_011540859.2 |
3142 |
Missense Mutation |
ATA,ATG |
I941M |
XP_011539161.1 |
XM_017000490.1 |
3142 |
Missense Mutation |
ATA,ATG |
I1305M |
XP_016855979.1 |
XM_017000491.1 |
3142 |
Missense Mutation |
ATA,ATG |
I1288M |
XP_016855980.1 |
XM_017000492.1 |
3142 |
Missense Mutation |
ATA,ATG |
I1011M |
XP_016855981.1 |
XM_017000493.1 |
3142 |
Missense Mutation |
ATA,ATG |
I775M |
XP_016855982.1 |
XM_017000494.1 |
3142 |
Intron |
|
|
XP_016855983.1 |
XM_017000495.1 |
3142 |
Intron |
|
|
XP_016855984.1 |
XM_017000496.1 |
3142 |
Intron |
|
|
XP_016855985.1 |
View Full Product Details