Product Details

SNP ID
rs150796066
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:43305065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGTTCGCACCAGGTCACGCTTCG[A/C]GGCTTCTCCAAGCCCTCGGACCTCG
Phenotype
MIM: 600222
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TIE1 PubMed Links

Gene Details

Gene
TIE1
Gene Name
tyrosine kinase with immunoglobulin like and EGF like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253357.1 367 Silent Mutation CGA,CGC R46R NP_001240286.1
NM_005424.4 367 Silent Mutation CGA,CGC R91R NP_005415.1
XM_005271163.1 367 Silent Mutation CGA,CGC R91R XP_005271220.1
XM_006710869.1 367 UTR 5 XP_006710932.1
XM_017002207.1 367 Silent Mutation CGA,CGC R91R XP_016857696.1

View Full Product Details