Product Details

SNP ID
rs150983499
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:53855313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAAGCTCACCCCAATCTGGATG[C/T]GGTCTCAAGCCCTCAAGCCAAAGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YIPF1 PubMed Links

Gene Details

Gene
YIPF1
Gene Name
Yip1 domain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018982.4 Intron NP_061855.1
XM_017001499.1 Intron XP_016856988.1
XM_017001500.1 Intron XP_016856989.1
XM_017001501.1 Intron XP_016856990.1

View Full Product Details