Product Details

SNP ID
rs151008086
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:151051786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTCTAGGACTAGGGCACAGAA[A/G]TAGAGCTTAAAATTTGGGGAGGATA
Phenotype
MIM: 611275 MIM: 604684
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BNIPL PubMed Links

Gene Details

Gene
BNIPL
Gene Name
BCL2 interacting protein like
There are no transcripts associated with this gene.

Gene
C1orf56
Gene Name
chromosome 1 open reading frame 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017860.3 2232 Intron NP_060330.2
Gene
CDC42SE1
Gene Name
CDC42 small effector 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038707.1 2232 UTR 3 NP_001033796.1
NM_020239.3 2232 UTR 3 NP_064624.1
XM_017001847.1 2232 UTR 3 XP_016857336.1
Gene
MLLT11
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11
There are no transcripts associated with this gene.

View Full Product Details