Product Details

SNP ID
rs151037445
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:34851283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCCAACAAAAATTCTTACCCCT[A/G]TTTTACAGGTGAGGATATTGAGGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM12 PubMed Links

Gene Details

Gene
SMIM12
Gene Name
small integral membrane protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164824.1 4865 UTR 3 NP_001158296.1
NM_001164825.1 4865 UTR 3 NP_001158297.1
NM_001320261.1 4865 UTR 3 NP_001307190.1
NM_138428.5 4865 UTR 3 NP_612437.3
XM_005270403.4 4865 Intron XP_005270460.1
XM_011540553.2 4865 Intron XP_011538855.1
XM_011540554.2 4865 Intron XP_011538856.2
XM_011540555.2 4865 Intron XP_011538857.1
XM_017000166.1 4865 Intron XP_016855655.1

View Full Product Details