Product Details

SNP ID
rs151334316
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:41481183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGACTTCCGGCTTGGGACTGCCC[C/T]GGCTTCAGTCCTACGTGAATAGCAT
Phenotype
MIM: 131241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EDN2 PubMed Links

Gene Details

Gene
EDN2
Gene Name
endothelin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302269.1 437 Intron NP_001289198.1
NM_001956.4 437 Missense Mutation NP_001947.1
XM_017000512.1 437 Intron XP_016856001.1

View Full Product Details