Product Details
- SNP ID
-
rs138201035
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:259140 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATACTGCAGGACATGTTGCCACT[C/G]GGGGGAGACAGCATTGTTCATGTGC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB132
PubMed Links
Gene Details
- Gene
- DEFB132
- Gene Name
- defensin beta 132
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207469.2 |
165 |
Missense Mutation |
TCG,TGG |
S41W |
NP_997352.1 |
View Full Product Details