Product Details

SNP ID
rs138360482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33677154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGGATCTGTGGTGAGGGATGA[A/G]GGGGGAGATGATGGTGGTGGTGACA
Phenotype
MIM: 189971 MIM: 612478
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTL10 PubMed Links

Gene Details

Gene
ACTL10
Gene Name
actin like 10
There are no transcripts associated with this gene.

Gene
E2F1
Gene Name
E2F transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005225.2 1157 Silent Mutation CCC,CCT P339P NP_005216.1
Gene
NECAB3
Gene Name
N-terminal EF-hand calcium binding protein 3
There are no transcripts associated with this gene.

View Full Product Details