Product Details

SNP ID
rs138739946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:4781517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATACGTACAGGGAAGACTATCTT[C/T]TGTGCTTTTCAGCCTCTCCCCAAGA
Phenotype
MIM: 609492
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RASSF2 PubMed Links

Gene Details

Gene
RASSF2
Gene Name
Ras association domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014737.2 3828 UTR 3 NP_055552.1
NM_170774.1 3828 UTR 3 NP_739580.1
XM_005260895.3 3828 UTR 3 XP_005260952.1
XM_011529410.1 3828 UTR 3 XP_011527712.1
XM_011529411.1 3828 UTR 3 XP_011527713.1
XM_011529412.2 3828 UTR 3 XP_011527714.1
XM_017028149.1 3828 UTR 3 XP_016883638.1
XM_017028150.1 3828 UTR 3 XP_016883639.1
XM_017028151.1 3828 UTR 3 XP_016883640.1
XM_017028152.1 3828 UTR 3 XP_016883641.1
XM_017028153.1 3828 UTR 3 XP_016883642.1

View Full Product Details