Product Details
- SNP ID
-
rs138988702
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23364606 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGAAGAGTGTGGATGGTACTAGG[A/T]CTAATGTCTACTTAAATGAAGTGCA
- Phenotype
-
MIM: 613842
MIM: 611270
MIM: 605811
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GZF1
PubMed Links
Gene Details
- Gene
- GZF1
- Gene Name
- GDNF inducible zinc finger protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001317012.1 |
322 |
Missense Mutation |
ACT,TCT |
T75S |
NP_001303941.1 |
NM_001317019.1 |
322 |
Missense Mutation |
ACT,TCT |
T75S |
NP_001303948.1 |
NM_022482.4 |
322 |
Missense Mutation |
ACT,TCT |
T75S |
NP_071927.1 |
XM_005260800.2 |
322 |
Missense Mutation |
ACT,TCT |
T75S |
XP_005260857.1 |
XM_005260802.3 |
322 |
Missense Mutation |
ACT,TCT |
T75S |
XP_005260859.1 |
XM_011529321.2 |
322 |
Missense Mutation |
ACT,TCT |
T75S |
XP_011527623.1 |
XM_011529322.1 |
322 |
Missense Mutation |
ACT,TCT |
T75S |
XP_011527624.1 |
XM_017028017.1 |
322 |
Missense Mutation |
ACT,TCT |
T75S |
XP_016883506.1 |
XM_017028018.1 |
322 |
Missense Mutation |
ACT,TCT |
T75S |
XP_016883507.1 |
XM_017028019.1 |
322 |
Missense Mutation |
ACT,TCT |
T75S |
XP_016883508.1 |
- Gene
- LINC01431
- Gene Name
- long intergenic non-protein coding RNA 1431
There are no transcripts associated with this gene.
- Gene
- NAPB
- Gene Name
- NSF attachment protein beta
There are no transcripts associated with this gene.
- Gene
- NXT1
- Gene Name
- nuclear transport factor 2 like export factor 1
There are no transcripts associated with this gene.
View Full Product Details