Product Details

SNP ID
rs140023782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35228178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCATGGAGCTCAGGGGGTCATA[C/T]GTGCACTGAAATTGCGTACAAATTT
Phenotype
MIM: 604871
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP24 PubMed Links

Gene Details

Gene
MMP24
Gene Name
matrix metallopeptidase 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006690.3 Intron NP_006681.1
XM_011528500.2 Intron XP_011526802.1
XM_017027597.1 Intron XP_016883086.1
XM_017027598.1 Intron XP_016883087.1
Gene
MT1P3
Gene Name
metallothionein 1 pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details