Product Details

SNP ID
rs140300779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50512458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCAAGCATGGTTAAAATTGTATT[C/T]AGCATGCAACTAGCATGGAGTGTGT
Phenotype
MIM: 176885
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPN1 PubMed Links

Gene Details

Gene
PTPN1
Gene Name
protein tyrosine phosphatase, non-receptor type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278618.1 Intron NP_001265547.1
NM_002827.3 Intron NP_002818.1

View Full Product Details