Product Details
- SNP ID
-
rs140341521
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45253041 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGCAGGATTTCCCACACATGCCCA[C/T]GCAACACTTCAAGTCACGCTTGCAC
- Phenotype
-
MIM: 107285
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLPI
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6032073] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLPI
- Gene Name
- secretory leukocyte peptidase inhibitor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003064.3 |
376 |
Missense Mutation |
ATG,GTG |
M119V |
NP_003055.1 |
View Full Product Details