Product Details

SNP ID
rs140902364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:24963818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCAGCTATCTGGGAGGGCTAAA[C/T]AGCCTGGAGGCTGAGTCTGCAGAGG
Phenotype
MIM: 615884 MIM: 603253
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APMAP PubMed Links

Gene Details

Gene
APMAP
Gene Name
adipocyte plasma membrane associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020531.2 1347 Missense Mutation ATT,GTT I416V NP_065392.1
XM_005260763.3 1347 UTR 3 XP_005260820.1
Gene
CST7
Gene Name
cystatin F
There are no transcripts associated with this gene.

View Full Product Details