Product Details

SNP ID
rs140926257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35003494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGTTGTGGAAGTTGTTGAGCAG[A/G]AAGCCGGGGTACTTCTTGCTGTGCT
Phenotype
MIM: 609928 MIM: 608430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYH7B PubMed Links

Gene Details

Gene
MYH7B
Gene Name
myosin heavy chain 7B
There are no transcripts associated with this gene.

Gene
TRPC4AP
Gene Name
transient receptor potential cation channel subfamily C member 4 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015638.2 1684 Silent Mutation TTC,TTT F724F NP_056453.1
NM_199368.1 1684 Silent Mutation TTC,TTT F716F NP_955400.1
XM_011528772.2 1684 Intron XP_011527074.1
XM_011528774.1 1684 Silent Mutation TTC,TTT F326F XP_011527076.1
XM_017027799.1 1684 Intron XP_016883288.1
XM_017027800.1 1684 Silent Mutation TTC,TTT F386F XP_016883289.1
XM_017027801.1 1684 Silent Mutation TTC,TTT F326F XP_016883290.1

View Full Product Details