Product Details

SNP ID
rs141039910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:9516024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATCACAGAACAGGCCGATATC[A/G]CATTGACCCGGGGAGCTGAGGTGAA
Phenotype
MIM: 614641
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAMP5 PubMed Links

Gene Details

Gene
LAMP5
Gene Name
lysosomal associated membrane protein family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199897.1 757 Intron NP_001186826.1
NM_012261.3 757 Missense Mutation ACA,GCA T88A NP_036393.1
Gene
LAMP5-AS1
Gene Name
LAMP5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details