Product Details

SNP ID
rs141382500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25007888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTCACTAGTGATGATCTTCCT[C/T]AGGAGCCGCCGCATGACCTTCCCAG
Phenotype
MIM: 614355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSS1 PubMed Links

Gene Details

Gene
ACSS1
Gene Name
acyl-CoA synthetase short-chain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252675.1 2024 Silent Mutation CTA,CTG L646L NP_001239604.1
NM_001252676.1 2024 Silent Mutation CTA,CTG L527L NP_001239605.1
NM_001252677.1 2024 Intron NP_001239606.1
NM_032501.3 2024 Silent Mutation CTA,CTG L648L NP_115890.2
XM_006723659.1 2024 Silent Mutation CTA,CTG L443L XP_006723722.1
XM_011529388.1 2024 Silent Mutation CTA,CTG L430L XP_011527690.1

View Full Product Details