Product Details

SNP ID
rs141912408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58996790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTTAGCCAGTCTTTCTGTTGCC[A/G]TTATTCCACAGCTGAGAGCAAGCAG
Phenotype
MIM: 603169 MIM: 605297
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTSZ PubMed Links

Gene Details

Gene
CTSZ
Gene Name
cathepsin Z
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001336.3 776 Missense Mutation ACG,ATG T217M NP_001327.2
Gene
NELFCD
Gene Name
negative elongation factor complex member C/D
There are no transcripts associated with this gene.

View Full Product Details