Product Details

SNP ID
rs141969034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33662379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATAGTTCCCACAAAAGGACCAAA[G/T]CACCCAAGCAGGCCCGCAAGGAGAG
Phenotype
MIM: 612478
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACTL10 PubMed Links

Gene Details

Gene
ACTL10
Gene Name
actin like 10
There are no transcripts associated with this gene.

Gene
C20orf144
Gene Name
chromosome 20 open reading frame 144
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080825.3 94 Missense Mutation GCA,TCA A12S NP_543015.1
Gene
NECAB3
Gene Name
N-terminal EF-hand calcium binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031231.3 94 Intron NP_112508.3
NM_031232.3 94 Intron NP_112509.3
XM_005260510.1 94 Intron XP_005260567.1
XM_011528991.1 94 Intron XP_011527293.1
XM_011528992.2 94 Intron XP_011527294.1
XM_017028015.1 94 Intron XP_016883504.1
XM_017028016.1 94 Intron XP_016883505.1

View Full Product Details