Product Details
- SNP ID
-
rs142084089
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:33360380 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAGCACATAGTCCCCAGGCTGGG[C/T]TCTGACCCTGAGCCCAGGGTTATTG
- Phenotype
-
MIM: 608200
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDK5RAP1
PubMed Links
Gene Details
- Gene
- CDK5RAP1
- Gene Name
- CDK5 regulatory subunit associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001278167.1 |
880 |
Missense Mutation |
ACC,GCC |
T551A |
NP_001265096.1 |
NM_001278168.1 |
880 |
Missense Mutation |
ACC,GCC |
T475A |
NP_001265097.1 |
NM_001278169.1 |
880 |
Missense Mutation |
ACC,GCC |
T525A |
NP_001265098.1 |
NM_016082.4 |
880 |
Missense Mutation |
ACC,GCC |
T553A |
NP_057166.4 |
NM_016408.3 |
880 |
Missense Mutation |
ACC,GCC |
T552A |
NP_057492.2 |
XM_011528856.2 |
880 |
Missense Mutation |
ACC,GCC |
T448A |
XP_011527158.1 |
XM_011528857.2 |
880 |
Missense Mutation |
ACC,GCC |
T442A |
XP_011527159.1 |
XM_011528858.1 |
880 |
Missense Mutation |
ACC,GCC |
T425A |
XP_011527160.1 |
XM_011528859.2 |
880 |
Missense Mutation |
ACC,GCC |
T425A |
XP_011527161.1 |
XM_011528860.1 |
880 |
Missense Mutation |
ACC,GCC |
T298A |
XP_011527162.1 |
XM_017027876.1 |
880 |
Missense Mutation |
ACC,GCC |
T298A |
XP_016883365.1 |
XM_017027877.1 |
880 |
Missense Mutation |
ACC,GCC |
T286A |
XP_016883366.1 |
View Full Product Details