Product Details

SNP ID
rs142799785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25075055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTTTAATTTTCCCAAGAGTATG[A/G]AACTAACATAGGATGAGAAAAATAG
Phenotype
MIM: 605020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VSX1 PubMed Links

Gene Details

Gene
VSX1
Gene Name
visual system homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256271.1 Intron NP_001243200.1
NM_001256272.1 Intron NP_001243201.1
NM_014588.5 Intron NP_055403.2
NM_199425.2 Intron NP_955457.1
XM_017027837.1 Intron XP_016883326.1
XM_017027838.1 Intron XP_016883327.1

View Full Product Details