Product Details

SNP ID
rs143394664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18293340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGACATATTTTCGTGGGTTCCA[G/T]CACATATTTTCGTGGGTTGGCTGGA
Phenotype
MIM: 604075
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 3523 Intron NP_001076799.1
NM_001282995.1 3523 Intron NP_001269924.1
NM_001282996.1 3523 Intron NP_001269925.1
NM_001282997.1 3523 Intron NP_001269926.1
NM_001282998.1 3523 Intron NP_001269927.1
NM_001282999.1 3523 Intron NP_001269928.1
NM_001283000.1 3523 Intron NP_001269929.1
NM_001283001.1 3523 Intron NP_001269930.1
NM_001283002.1 3523 Intron NP_001269931.1
NM_001283003.1 3523 Intron NP_001269932.1
NM_001283004.1 3523 Intron NP_001269933.1
NM_001283005.1 3523 Intron NP_001269934.1
NM_001283006.1 3523 Intron NP_001269935.1
NM_001283007.1 3523 Intron NP_001269936.1
NM_001283008.1 3523 Intron NP_001269937.1
NM_003434.5 3523 Intron NP_003425.2
XM_005260819.1 3523 Intron XP_005260876.1
XM_005260820.4 3523 Intron XP_005260877.1
XM_011529336.1 3523 Intron XP_011527638.1
XM_011529337.1 3523 Intron XP_011527639.1
XM_011529338.2 3523 Intron XP_011527640.1
XM_011529339.1 3523 Intron XP_011527641.1
XM_011529340.1 3523 Intron XP_011527642.1
XM_017028041.1 3523 UTR 5 XP_016883530.1
XM_017028042.1 3523 Intron XP_016883531.1
XM_017028043.1 3523 UTR 5 XP_016883532.1
XM_017028044.1 3523 UTR 5 XP_016883533.1
XM_017028045.1 3523 UTR 5 XP_016883534.1
XM_017028046.1 3523 UTR 5 XP_016883535.1
XM_017028047.1 3523 Intron XP_016883536.1
XM_017028048.1 3523 Intron XP_016883537.1
XM_017028049.1 3523 UTR 5 XP_016883538.1
XM_017028050.1 3523 Intron XP_016883539.1
XM_017028051.1 3523 Intron XP_016883540.1
XM_017028052.1 3523 Intron XP_016883541.1
XM_017028053.1 3523 Intron XP_016883542.1
XM_017028054.1 3523 Intron XP_016883543.1
XM_017028055.1 3523 Intron XP_016883544.1
XM_017028056.1 3523 Intron XP_016883545.1
XM_017028057.1 3523 Intron XP_016883546.1
XM_017028058.1 3523 Intron XP_016883547.1

View Full Product Details