Product Details
- SNP ID
-
rs143625916
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23047836 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGCAGCACTACCTCCTTGGAAG[C/G]GGCCGCGCACTTGTACTCCATCTTG
- Phenotype
-
MIM: 188040
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
THBD
PubMed Links
Gene Details
- Gene
- THBD
- Gene Name
- thrombomodulin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000361.2 |
1829 |
Missense Mutation |
CCT,GCT |
P557A |
NP_000352.1 |
View Full Product Details