Product Details

SNP ID
rs143638505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46062721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATTGGCCGTCACTGTGCCACTA[C/T]TGAAGAGGCTGAGGATTTTGGCCTG
Phenotype
MIM: 616825 MIM: 606726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCOA5 PubMed Links

Gene Details

Gene
NCOA5
Gene Name
nuclear receptor coactivator 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020967.2 1450 Missense Mutation AAT,AGT N440S NP_066018.1
XM_005260474.3 1450 Missense Mutation AAT,AGT N335S XP_005260531.1
XM_011528951.2 1450 UTR 3 XP_011527253.1
XM_017027988.1 1450 UTR 3 XP_016883477.1
Gene
SLC12A5
Gene Name
solute carrier family 12 member 5
There are no transcripts associated with this gene.

View Full Product Details