Product Details

SNP ID
rs143932195
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63555836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTCCTGACCTCGGACCTGAGTC[C/T]GCACCTGCTCAAGCGCCACCACGCC
Phenotype
MIM: 611265
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FNDC11 PubMed Links
Additional Information
For this assay, SNP(s) [rs3746348] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FNDC11
Gene Name
fibronectin type III domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319152.1 249 Missense Mutation CCG,CTG P58L NP_001306081.1
NM_001319153.1 249 Missense Mutation CCG,CTG P58L NP_001306082.1
NM_024059.3 249 Missense Mutation CCG,CTG P58L NP_076964.1
XM_011529041.2 249 Missense Mutation CCG,CTG P58L XP_011527343.1
XM_017028060.1 249 Missense Mutation CCG,CTG P63L XP_016883549.1
Gene
HELZ2
Gene Name
helicase with zinc finger 2
There are no transcripts associated with this gene.

Gene
SRMS
Gene Name
src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites
There are no transcripts associated with this gene.

View Full Product Details