Product Details
- SNP ID
-
rs144051050
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:41345832 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTACGTGGGGCAGCTGGCGGAGA[C/T]GGTGTTTGGGACGGTGAAGGAGCTG
- Phenotype
-
MIM: 605520
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LPIN3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16985673] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LPIN3
- Gene Name
- lipin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301860.1 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
NP_001288789.1 |
XM_005260516.2 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_005260573.1 |
XM_006723863.2 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_006723926.1 |
XM_011528996.1 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_011527298.1 |
XM_011528997.2 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_011527299.1 |
XM_011528998.2 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_011527300.1 |
XM_011528999.2 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_011527301.1 |
XM_011529000.1 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_011527302.1 |
XM_011529001.1 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_011527303.1 |
XM_011529002.2 |
185 |
Intron |
|
|
XP_011527304.1 |
XM_011529003.1 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_011527305.1 |
XM_011529004.1 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_011527306.1 |
XM_011529005.2 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_011527307.1 |
XM_011529006.2 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_011527308.1 |
XM_017028020.1 |
185 |
Missense Mutation |
ACG,ATG |
T10M |
XP_016883509.1 |
View Full Product Details