Product Details

SNP ID
rs144360449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57561542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGTGTCAGCCTGATCACATCC[A/G]CATCTGTGACGGCTCTGAGGAGGAG
Phenotype
MIM: 614168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCK1 PubMed Links

Gene Details

Gene
PCK1
Gene Name
phosphoenolpyruvate carboxykinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002591.3 294 Missense Mutation CAC,CGC H44R NP_002582.3
XM_011528839.1 294 UTR 5 XP_011527141.1

View Full Product Details