Product Details

SNP ID
rs144903215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37301293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGACCTAGACTTCTCACCTCC[A/G]GAGGTGCCCGAGCCCACTTTCCTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MANBAL PubMed Links

Gene Details

Gene
MANBAL
Gene Name
mannosidase beta like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003897.1 1300 Silent Mutation CCA,CCG P10P NP_001003897.1
NM_022077.3 1300 Silent Mutation CCA,CCG P10P NP_071360.1
XM_017028006.1 1300 Silent Mutation CCA,CCG P10P XP_016883495.1
XM_017028007.1 1300 Silent Mutation CCA,CCG P10P XP_016883496.1

View Full Product Details