Product Details

SNP ID
rs145458910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44890870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAGAGACACTTGCATTGCAGT[A/G]AACTGTGATAAGGGCCATTTTAGAA
Phenotype
MIM: 601289
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YWHAB PubMed Links

Gene Details

Gene
YWHAB
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003404.4 Intron NP_003395.1
NM_139323.3 Intron NP_647539.1
XM_017028039.1 Intron XP_016883528.1
XM_017028040.1 Intron XP_016883529.1

View Full Product Details