Product Details
- SNP ID
-
rs145488639
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:259108 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGTGGGTCAAAATGTGTGAGTAA[C/T]ACCCCAGGATACTGCAGGACATGTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB132
PubMed Links
Gene Details
- Gene
- DEFB132
- Gene Name
- defensin beta 132
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207469.2 |
133 |
Silent Mutation |
AAC,AAT |
N30N |
NP_997352.1 |
View Full Product Details