Product Details

SNP ID
rs145700972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3796718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACCCAGACCATGCACGACCTC[A/G]CCGGGCTCGGCAGGTAGGACACCCC
Phenotype
MIM: 116949
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC25B PubMed Links

Gene Details

Gene
CDC25B
Gene Name
cell division cycle 25B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287516.1 980 Intron NP_001274445.1
NM_001287517.1 980 Intron NP_001274446.1
NM_001287518.1 980 Intron NP_001274447.1
NM_001287519.1 980 Intron NP_001274448.1
NM_001287520.1 980 Intron NP_001274449.1
NM_001287522.1 980 Intron NP_001274451.1
NM_001287524.1 980 Intron NP_001274453.1
NM_004358.4 980 Missense Mutation ACC,GCC T63A NP_004349.1
NM_021872.3 980 Missense Mutation ACC,GCC T63A NP_068658.1
NM_021873.3 980 Missense Mutation ACC,GCC T63A NP_068659.1

View Full Product Details