Product Details
- SNP ID
-
rs146373315
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:3669302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGAATCCAGGTCCCTGCCTACCTT[G/T]GGGGTCAGGCGAGACTGCTGGCAGA
- Phenotype
-
MIM: 607114
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADAM33
PubMed Links
Gene Details
- Gene
- ADAM33
- Gene Name
- ADAM metallopeptidase domain 33
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282447.2 |
2466 |
Missense Mutation |
AAA,CAA |
K801Q |
NP_001269376.1 |
NM_025220.4 |
2466 |
Missense Mutation |
AAA,CAA |
K801Q |
NP_079496.1 |
NM_153202.3 |
2466 |
Missense Mutation |
AAA,CAA |
K775Q |
NP_694882.1 |
XM_005260843.1 |
2466 |
Missense Mutation |
AAA,CAA |
K814Q |
XP_005260900.1 |
XM_006723639.1 |
2466 |
Missense Mutation |
AAA,CAA |
K814Q |
XP_006723702.1 |
XM_006723640.1 |
2466 |
Missense Mutation |
AAA,CAA |
K811Q |
XP_006723703.1 |
XM_006723644.2 |
2466 |
Intron |
|
|
XP_006723707.1 |
XM_011529366.1 |
2466 |
Missense Mutation |
AAA,CAA |
K813Q |
XP_011527668.1 |
XM_011529367.1 |
2466 |
Missense Mutation |
AAA,CAA |
K800Q |
XP_011527669.1 |
XM_011529368.1 |
2466 |
Missense Mutation |
AAA,CAA |
K788Q |
XP_011527670.1 |
XM_011529370.1 |
2466 |
Intron |
|
|
XP_011527672.1 |
XM_011529371.1 |
2466 |
Intron |
|
|
XP_011527673.1 |
XM_011529373.1 |
2466 |
Missense Mutation |
AAA,CAA |
K480Q |
XP_011527675.1 |
XM_017028080.1 |
2466 |
Intron |
|
|
XP_016883569.1 |
XM_017028081.1 |
2466 |
Intron |
|
|
XP_016883570.1 |
XM_017028082.1 |
2466 |
Intron |
|
|
XP_016883571.1 |
XM_017028083.1 |
2466 |
Intron |
|
|
XP_016883572.1 |
- Gene
- GFRA4
- Gene Name
- GDNF family receptor alpha 4
There are no transcripts associated with this gene.
View Full Product Details