Product Details
- SNP ID
-
rs146671297
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:33359046 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCTGAGGCCATCCTCTCAGGTCA[C/G]CAATATGCAGAAGAGTCCCTCAGAG
- Phenotype
-
MIM: 608200
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CDK5RAP1
PubMed Links
Gene Details
- Gene
- CDK5RAP1
- Gene Name
- CDK5 regulatory subunit associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001278167.1 |
987 |
Missense Mutation |
TGC,TGG |
C586W |
NP_001265096.1 |
NM_001278168.1 |
987 |
Missense Mutation |
TGC,TGG |
C510W |
NP_001265097.1 |
NM_001278169.1 |
987 |
Missense Mutation |
TGC,TGG |
C560W |
NP_001265098.1 |
NM_016082.4 |
987 |
Missense Mutation |
TGC,TGG |
C588W |
NP_057166.4 |
NM_016408.3 |
987 |
Missense Mutation |
TGC,TGG |
C587W |
NP_057492.2 |
XM_011528856.2 |
987 |
Missense Mutation |
TGC,TGG |
C483W |
XP_011527158.1 |
XM_011528857.2 |
987 |
Missense Mutation |
TGC,TGG |
C477W |
XP_011527159.1 |
XM_011528858.1 |
987 |
Missense Mutation |
TGC,TGG |
C460W |
XP_011527160.1 |
XM_011528859.2 |
987 |
Missense Mutation |
TGC,TGG |
C460W |
XP_011527161.1 |
XM_011528860.1 |
987 |
Missense Mutation |
TGC,TGG |
C333W |
XP_011527162.1 |
XM_017027876.1 |
987 |
Missense Mutation |
TGC,TGG |
C333W |
XP_016883365.1 |
XM_017027877.1 |
987 |
Missense Mutation |
TGC,TGG |
C321W |
XP_016883366.1 |
View Full Product Details