Product Details

SNP ID
rs147320694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:22563108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTATATGTATGGGATCAACATA[C/T]AATTCCAGAGATAAAAGCTGAAAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00261 PubMed Links

Gene Details

Gene
LINC00261
Gene Name
long intergenic non-protein coding RNA 261
There are no transcripts associated with this gene.

View Full Product Details