Product Details

SNP ID
rs147547175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2815211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCCGGCTCTGCTTACCCAGACC[C/T]GAAGCACGTGAGGATCCGATCCCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf141 PubMed Links

Gene Details

Gene
C20orf141
Gene Name
chromosome 20 open reading frame 141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256538.1 201 Silent Mutation CCC,CCT P9P NP_001243467.1
NM_080739.2 201 Silent Mutation CCC,CCT P9P NP_542777.1
Gene
TMEM239
Gene Name
transmembrane protein 239
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167670.2 201 Intron NP_001161142.1
NM_001318207.1 201 Intron NP_001305136.1
XM_011529128.2 201 Intron XP_011527430.1
XM_011529129.2 201 Intron XP_011527431.1

View Full Product Details