Product Details

SNP ID
rs147559982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31477883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAGAAGACACCACACTGGTGGT[C/T]GTGGACACCTGGGAGGCCGAGAAAC
Phenotype
MIM: 610388
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEFB124 PubMed Links

Gene Details

Gene
DEFB124
Gene Name
defensin beta 124
There are no transcripts associated with this gene.

Gene
LINC00028
Gene Name
long intergenic non-protein coding RNA 28
There are no transcripts associated with this gene.

Gene
REM1
Gene Name
RRAD and GEM like GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014012.5 712 Silent Mutation GTC,GTT V132V NP_054731.2
XM_005260404.1 712 Silent Mutation GTC,GTT V140V XP_005260461.1
XM_011528795.1 712 Silent Mutation GTC,GTT V138V XP_011527097.1
XM_017027833.1 712 Silent Mutation GTC,GTT V140V XP_016883322.1

View Full Product Details