Product Details

SNP ID
rs148866137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45899641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTACCATTGAGCATGGGCATCAC[C/G]CCAATCTGCAGCATGGTCTTCAGAG
Phenotype
MIM: 613111 MIM: 608597 MIM: 172425
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTSA PubMed Links

Gene Details

Gene
CTSA
Gene Name
cathepsin A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000308.3 1275 Intron NP_000299.2
NM_001127695.2 1275 Intron NP_001121167.1
NM_001167594.2 1275 Intron NP_001161066.1
Gene
NEURL2
Gene Name
neuralized E3 ubiquitin protein ligase 2
There are no transcripts associated with this gene.

Gene
PLTP
Gene Name
phospholipid transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242920.1 1275 Silent Mutation GGC,GGG G326G NP_001229849.1
NM_001242921.1 1275 Silent Mutation GGC,GGG G333G NP_001229850.1
NM_006227.3 1275 Silent Mutation GGC,GGG G421G NP_006218.1
NM_182676.2 1275 Silent Mutation GGC,GGG G369G NP_872617.1

View Full Product Details