Product Details

SNP ID
rs149005015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:54477827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTTTATGCCAAATTGTGACACA[C/T]GGTAATTTATTTGAAAAACTGGTTA
Phenotype
MIM: 608334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOK5 PubMed Links

Gene Details

Gene
DOK5
Gene Name
docking protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294161.1 Intron NP_001281090.1
NM_018431.4 Intron NP_060901.2
NM_177959.2 Intron NP_808874.1
XM_011528904.1 Intron XP_011527206.1

View Full Product Details